Learn More
Medchemexpress LLC AC-261066 | 870773-76-5 | 99.3% | 353.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10853250MG
Encompass_Preferred
AC-261066 is a potent, orally available, and isoform-selective retinoic acid beta2 (RARbeta2) receptor agonist, with a pEC50 of 8.0.
  • Exhibits good oral bioavailability in rats (F oral = 52%).
Catalog No. 50-003-46672