Learn More
Medchemexpress LLC AC-263093 (free base) | 849459-86-5 | 99.8% | 319.98 g/mol | C8H8Br2N4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12468010MG
Encompass_Preferred
AC-263093 (free base) is a small-molecule research compound reported to activate neuropeptide FF receptor 2 (NPFFR2). Provided as a free base for laboratory use, it is suitable for preclinical receptor pharmacology and biochemical assays. Key identifiers include CAS 849459-86-5, molecular formula C8H8Br2N4, molecular weight 319.98 g/mol, and reported high purity.
  • Activates NPFFR2 for receptor pharmacology studies.
  • Provided as a free base for formulation flexibility.
  • High reported purity supports reproducible results.
  • Available in small-scale quantities for preclinical work.
  • Batch-level documentation available for verification.
Catalog No. 50-002-36356