Learn More
Medchemexpress LLC Ac-Ala-OH | 97-69-8 | MFCD00037238 | 131.13 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW004066500MG
Encompass_Preferred
Ac-Ala-OH is recognized as an endogenous metabolite that has been shown to inhibit acylaminoacyl peptide hydrolase (APEH). This compound has potential applications in breast cancer research.
  • Endogenous metabolite
  • Inhibits acylaminoacyl peptide hydrolase (APEH)
  • Potential use in breast cancer research
Catalog No. 50-004-47511