Learn More
Medchemexpress LLC Ac-Arg-Gly-Lys(Ac)-AMC | 660846-97-9 | >99.8% | 600.67 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP24625MG
Encompass_Preferred
Ac-Arg-Gly-Lys(Ac)-AMC is a substrate for Histone Deacetylase (HDAC) research, intended for laboratory use only. This white to off-white solid has a purity of >99.8% and a molecular weight of 600.67. It is soluble in water at concentrations of ≥ 50 mg/mL (83.24 mM).
  • Substrate for HDAC research
  • High purity of >99.8%
  • Soluble in water (≥ 50 mg/mL)
  • Suitable for in vitro studies
  • Specific storage conditions for powder and in solvent
Catalog No. 50-004-23737