Learn More
Medchemexpress LLC Ac-Arg-Gly-Lys(Ac)-AMC | 660846-97-9 | 99.9% | 600.67 | 10 MG

Supplier: Medchemexpress LLC HYP246210MG
Ac-Arg-Gly-Lys(Ac)-AMC is a chemical reagent used in research applications. It functions as a substrate for Histone Deacetylase (HDAC) enzymes, which play a critical role in epigenetic regulation.
- Purity: 99.9%
- Molecular weight: 600.67
- Physical form: Solid
- Color: White to off-white
- Storage conditions: Sealed, away from moisture and light, under nitrogen. Powder: -80°C for 2 years or -20°C for 1 year.
- Solubility: ≥ 50 mg/mL (83.24 mM) in H2O.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.