Learn More
Bachem Ac-Arg-Leu-Arg-AMC
SDP
Supplier:  Bachem 4053458.0005
Encompass_Preferred
Ac-Arg-Leu-Arg-AMC Trifluoroacetate, 929903-87-7, C30H46N10O6, Mr 642.76, 5mg. Ac-RLR-AMC, fluorogenic substrate for assaying the trypsin-like activity of purified proteasomes (Km = 78 μM).
Catalog No. 50-260-360