Learn More
Bachem Ac-Arg-pNA · HCl
SDP
Supplier:  Bachem 4004007.025
Encompass_Preferred
Ac-Arg-pNA · HCl, 40127-26-2, C14H20N6O4 · HCl, Mr 372.81, 250mg. L-AAPA. Chromogenic substrate for trypsin and papain.
Catalog No. NC1632950