Learn More
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin, MP Biomedicals™
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin is a fluorescent substrate.
Supplier:  MP Biomedicals, Inc 03AFC13810
 View more versions of this product
Catalog No. 03AFC13810
Add to Cart
Description
Description
  • Ac-DEVD-AFC is a sensitive fluorescent substrate for the assay of caspase-3 and CPP-32 activity.
  • The trifluoromethyl substituent of the fluorophore improves the membrane permeability of the substrate.
  • Protein or Enzyme Type: Activators, Inhibitors and Substrates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lyophilized
10 mg
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.