Learn More
Cayman Chemical Ac-CalpastatIn184-210 100ug
SDP
Supplier:  Cayman Chemical 16501100 UG
Encompass_Preferred
An acetylated synthetic peptide from human calpastatin that strongly inhibits both calpains I and II but not papain or trypsin
Catalog No. 50-296-8242