Learn More
Medchemexpress LLC Ac-Cys Bzl -OH 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW101367
Encompass_Preferred
Ac-Cys(Bzl)-OH (N-Acetyl-S-benzyl-L-cysteine) is a toluene metabolite Ac-Cys(Bzl)-OH in urine samples is a good marker of occupational toluene exposure
Catalog No. 50-002-05745