Learn More
Medchemexpress LLC Ac-D-Ala-OH-d4 | 2708343-30-8 | 99.0% | 135.15 | 10 MG

Supplier: Medchemexpress LLC HYW012713S10MG
Ac-D-Ala-OH-d4 is a laboratory chemical primarily used for research purposes and in the manufacture of substances. It has not been fully validated for medical applications.
- Appears as a white to off-white solid
- Purity of 99.0%
- Isotopic enrichment of 99.40%
- Stable for 3 years as powder at -20°C
- Stable for 2 years as powder at 4°C
- Stable for 6 months in solvent at -80°C
- Stable for 1 month in solvent at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.