Learn More
Medchemexpress LLC Ac-(d-Arg)-CEH-(d-Phe)-RWC-NH2 | 819048-44-7 | 100.0% | 1175.35 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP47641MG
Encompass_Preferred
Ac-(d-Arg)-CEH-(d-Phe)-RWC-NH2 is a Melanocortin Receptor 4 (MC4R) Agonist, intended for research use. This high-purity compound is provided as a solid, offering stability under recommended storage conditions for various laboratory applications.
  • High purity of 99.9%
  • Identified by CAS number 819048-44-7
  • Molecular weight of 1175.35
  • Stable as a powder at -80°C for up to 2 years
Catalog No. 50-003-96684