Learn More
Apexbio Technology LLC Ac-DEVD-AFC 201608-14-2 10mg
SDP
Supplier:  Apexbio Technology LLC C552410
Encompass_Preferred
Ac-DEVD-AFC (CAS 201608-14-2) is a fluorogenic substrate targeting active caspase-3 It is designed to measure the enzymatic activity of caspase-3 thereby enabling the detection and quantification of apoptotic processes Ac-DEVD-AFC exerts its biological activity primarily through specific cleavage by active caspase-3 resulting in hydrolysis of the peptide bond and release of the fluorescent moiety AFC (7-amino-4-trifluoromethylcoumarin) Based on these properties Ac-DEVD-AFC holds research potential in the detection and quantification of caspase-3 mediated apoptosis in cell lysates tissue extracts and biochemical assays
Catalog No. 50-000-91575