Learn More
Selleck Chemical LLC Ac-DEVD-CHO
SDP
Supplier:  Selleck Chemical LLC S7901100MG
Encompass_Preferred
Ac-DEVD-CHO (Caspase-3 Inhibitor I N-Ac-Asp-Glu-Val-Asp-CHO) is a potent aldehyde inhibitor of Group II caspases with Ki values of 0 2 nM and 0 3 nM for for caspase-3 and caspase-7 respectively Weak inhibition for caspase-2
Catalog No. 50-000-00559