Learn More
Medchemexpress LLC Ac-DEVD-CHO | 169332-60-9 | 99.8% | 502.47 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP1001
Ac-DEVD-CHO is a potent Caspase-3 inhibitor with a Ki value of 230 pM. It also shows inhibitory activity against Caspase-8, Caspase-7, Caspase-10, Caspase-1, Caspase-6, Caspase-9, Caspase-4, Caspase-5, and Caspase-2. This product is for research use only and not sold to patients.
- Inhibits Caspase-3 with a Ki of 0.23 nM
- Inhibits Caspase-8 (0.92 nM), Caspase-7 (1.6 nM), Caspase-10 (12 nM), Caspase-1 (18 nM), Caspase-6 (31 nM), Caspase-9 (60 nM), Caspase-4 (132 nM), Caspase-5 (205 nM), and Caspase-2 (1710 nM)
- Appears as a white to off-white solid
- Soluble in H2O at ≥ 50 mg/mL (99.51 mM)
- Powder storage: -80°C for 2 years or -20°C for 1 year
- In solvent storage: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light, under nitrogen)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.