Learn More
Medchemexpress LLC Ac-DEVD-CHO | 169332-60-9 | 99.8% | 502.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP10015MG
Encompass_Preferred
Ac-DEVD-CHO is a potent Caspase-3 inhibitor with a Ki value of 230 pM. This product is widely used in research to study the role of Caspase-3 in various cellular processes, including apoptosis. It can prevent SLNT-induced apoptosis, reduce apoptosis rates after hypericin-mediated PDT treatment, and partially block SIN-induced apoptosis. It is suitable for applications in cell assays and animal administration protocols to investigate its inhibitory effects on caspases and apoptosis.
  • Inhibits Caspase-3 with a Ki value of 230 pM
  • Prevents SLNT-induced apoptosis
  • Decreases apoptosis rates after hypericin-mediated PDT treatment
  • Partially blocks SIN-induced apoptosis and reduces apoptotic nuclei
  • Suitable for cell assays and animal administration protocols
Catalog No. 50-004-25136