Learn More
TARGETMOL CHEMICALS INC Ac-DEVD-CHO 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ01462MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Ac-DEVD-CHO is a specific Caspase-3 inhibitor (Ki 230 pM) with inhibitory effects on SLNT-induced apoptosis. Purity 98.69%
Catalog No. 50-287-9419