Learn More
Enzo Life Sciences Ac-DEVD-CHO (5mg). CAS: 169332-60-9
SDP
Supplier:  Enzo Life Sciences ALX260030M005
Encompass_Preferred
Highly specific, potent and reversible inhibitor of caspase-3. Ki<1nM for purified caspase-3. IC50=0.2nM for PARP cleavage in cultured human osteosarcoma cell extracts. In contrast, the caspase-1 (ICE) specific inhibitor YVAD-CHO has a Ki=12µM for caspase-3 and IC50>10µM for PARP cleavage in osteosarcoma cell extracts. The DEVD sequence is based on PARP cleavage site Asp-216 for caspase-3. Alternative name: Caspase-3 inhibitor (aldehyde), Caspase-7 inhibitor (aldehyde). Formula: C20H30N4O11. MW: 502.5. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in distilled water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8241