Learn More
Cayman Chemical AC-DEVD-PNA 5MG
SDP
Supplier:  Cayman Chemical 144605 MG
Encompass_Preferred
A para-nitro aniline chromophore cleaved by caspases (Km = 18, 11, 32, 180, and 12 µM for caspases-1, -3, -4, -6, and -7, respectively); not cleaved by caspase-2; monitored colorimetrically at 405 nm
Catalog No. 50-297-0398