Learn More
Medchemexpress LLC Ac-exatecan | 2922852-48-8 | 99.0% | 477.48 | C26H24FN3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY7949010MG
Encompass_Preferred
Ac-Exatecan is an acetylated derivative of exatecan that acts as a DNA topoisomerase I inhibitor and is used as an antibody-drug conjugate (ADC) payload in research. It shows antitumor activity and retains activity against P-glycoprotein-mediated multidrug-resistant cells. Intended for research use only; not for human or veterinary use.
  • Acetylated exatecan derivative and topoisomerase I inhibitor.
  • High purity (reported 99.02%); enantiomeric excess 100%.
  • Molecular weight 477.48; formula C26H24FN3O5.
  • Solid, off-white to light yellow appearance.
  • Store protected from light at 4°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Suitable as an ADC payload and for cancer research applications.
Catalog No. 50-002-98772