Learn More
Medchemexpress LLC Ac-feid-cmk (TFA) | 97.1% | 711.08 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP3434A5MG
Encompass_Preferred
Ac-FEID-CMK TFA is a potent zebrafish-specific GSDMEb-derived peptide inhibitor. It inhibits pyroptosis and attenuates septic acute kidney injury (AKI) in vivo, and can suppress the caspy2-mediated noncanonical inflammasome pathway.
  • Attenuates mortality during septic shock
  • Reduces kidney injury during septic shock
  • Inhibits pyroptosis
  • Suppresses caspy2-mediated noncanonical inflammasome pathway
Catalog No. 50-003-97674