Learn More
Medchemexpress LLC HY-111675 1mg Medchemexpress, Ac-FLTD-CMK CAS:2376255-48-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1116751MG
Encompass_Preferred
Medchemexpress, HY-111675 1mg Ac-FLTD-CMK CAS:2376255-48-8 Ac-FLTD-CMK, a gasdermin D (GSDMD)-derived inhibitor, is a specific inflammatory caspases inhibitor. Ac-FLTD-CMK is effective against caspases-1 (IC50 of 46.7 nM), caspases-4 (IC50 of 1.49 μM), caspases-5 (IC50 of 329 nM), and caspases-11 , but not the apoptotic caspases such as caspase-3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0801