Learn More
CPC Scientific Ac-Glu-Ser-Met-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP055A
Encompass
Peptide aldehyde that corresponds to one of the cleavage sites of the inactive 32 kD caspase-3 precursor (amino acids 25-28). In a concentration range of 2.5-5 ?M, it blocks the formation of the p17 subunit and concomitantly induces the accumulation of the p20 peptide.ONE-LETTER SEQUENCE: Ac-ESMD-CHOMOLECULAR FORMULA: C19H30N4O10S1MOLECULAR WEIGHT:506.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [191338-87-1], RESEARCH AREA: ApoptosisREFERENCES: Z. Han et al., J. Biol. Chem. , 272, 13432 (1997)
Catalog No. 50-210-9016