Learn More
Medchemexpress LLC Ac-{Gly(N-me)}-Sar-Sar-Sar-Sar-Sar-Sar-Sar-Sar-Sar | 2857963-60-9 | 99.5% | 770.83 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP4166100MG
Encompass_Preferred
Ac-{Gly(N-me)}-Sar-Sar-Sar-Sar-Sar-Sar-Sar-Sar-Sar is a decapeptide used for the preparation of antibody-drug conjugates.
  • Conjugates hydrophobic camptothecin-based payloads to antibodies.
  • Significantly enhances conjugate solubility and stability.
  • Reduces aggregation at high drug-to-antibody ratios (DAR).
  • Facilitates release of native, active drug payload upon cleavage.
Catalog No. 50-003-98195