Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-IEPD-AFC | 1135417-31-0 | 99.97% | 725.67 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP10925MG

Encompass_Preferred

Ac-IEPD-AFC is a substrate of Granzyme B. It has a Km value of 585 μM for human granzyme B in assay buffer.

  • Substrate for Granzyme B
  • Km value for human granzyme B in assay buffer is 585 μM
  • Purity of 99.97%
  • White to off-white solid appearance
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-24843


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.