Learn More
Medchemexpress LLC Ac-IEPD-AMC | 216757-33-4 | 98.0% | 671.69 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP4417A1MG
Encompass_Preferred
Ac-IEPD-AMC TFA is a fluorescent substrate used to measure protease activity. It undergoes hydrolysis to release the fluorescent product 7-amino-4-methylcoumarin (AMC), which fluoresces under UV light irradiation and emits fluorescent signals.
  • Measures protease activity
  • Releases 7-amino-4-methylcoumarin (AMC) upon hydrolysis
  • Emits fluorescent signals under UV light
  • Functions as a fluorogenic substrate
  • Categorized as a fluorescent dye, peptide, and enzyme substrate
Catalog No. 50-003-97934