Learn More
Medchemexpress LLC Ac-IEPD-AMC TFA | 98.01% | 671.69 (free acid) | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP4417A5MG
Encompass_Preferred
Ac-IEPD-AMC TFA is a fluorescent substrate utilized to assess protease activity. It undergoes hydrolysis, releasing the fluorescent product 7-amino-4-methylcoumarin (AMC), which fluoresces under UV light and emits fluorescent signals. This product is intended for research use only.
  • Fluorescent substrate for protease activity
  • Undergoes hydrolysis to release AMC
  • AMC fluoresces under UV light
  • Inhibitors and substrates classification
  • Fluorescent dye
Catalog No. 50-003-96813