Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-IEPD-CHO | 352520-90-2 | 99.21% | 498.53 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP101495MG

Encompass_Preferred

Ac-IEPD-CHO is a reversible granzyme B inhibitor with a Ki of 80 nM. It is also a caspase-8 inhibitor. In vitro, Ac-IEPD-CHO (35-50 μg/mL, 1 h pretreatment followed by 16 h) diminishes Th17-polarized cells induced MO3.13 cell death.

  • Reversible granzyme B inhibitor with a Ki of 80 nM.
  • Acts as a caspase-8 inhibitor.
  • Diminishes Th17-polarized cells induced MO3.13 cell death in vitro.
  • Powder storage: -80°C for 2 years or -20°C for 1 year in sealed storage, away from moisture.
  • In solvent storage: -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture.
  • Recommended shipping at room temperature in continental US, may vary elsewhere.
  • For research use only.

Catalog No. 50-004-22588


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.