Learn More
Medchemexpress LLC Ac-IETD-CHO | 191338-86-0 | MFCD01318858 | 98.6% | 502.52 | C21H34N4O10 | 5 MG

Supplier: Medchemexpress LLC HY1139635MG
Ac-IETD-CHO is a tetrapeptide aldehyde (Ac-Ile-Glu-Thr-Asp-CHO) that reversibly inhibits granzyme B and caspase-8; it is used in research to block caspase-8-dependent apoptosis and related cellular processes.
- Purity 98.6% (HPLC).
- Molecular weight 502.52 Da.
- Chemical formula C21H34N4O10.
- Sequence Ac-Ile-Glu-Thr-Asp-CHO.
- Soluble in DMSO up to 100 mg/mL; in vivo formulations reported at ≥2.5 mg/mL.
- Storage powder -80°C (2 years) or -20°C (1 year); keep sealed and dry.
- Available pack size 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.