Learn More
Apexbio Technology LLC Ac-LEHD-AFC 210345-03-2 10mg
SDP
Supplier:  Apexbio Technology LLC A441910
Encompass_Preferred
Ac-LEHD-AFC (CAS 210345-03-2) is a fluorogenic peptide substrate specifically recognized and cleaved by caspase-9 Upon proteolytic cleavage by caspase-9 the AFC (7-amino-4-trifluoromethylcoumarin) moiety is released generating a fluorescent signal detectable with excitation at 400 nm and emission at 505 nm This property enables quantitative measurement of caspase-9 activity in apoptotic pathways and related cellular studies Ac-LEHD-AFC is widely utilized in research applications focused on elucidating mechanisms of apoptosis and assessing caspase-9 function in various biochemical and cell-based assays
Catalog No. 50-000-94767