Learn More
Medchemexpress LLC Ac-Leu-Arg-AMC | 929621-79-4 | 98.2% | 486.56 | 10 MG

Supplier: Medchemexpress LLC HYP144810MG
Ac-Leu-Arg-AMC is a fluorogenic peptide substrate.
- Molecular weight: 486.56
- Purity: 98.2%
- Appearance: White to off-white solid
- Emission (Em): 480
- Excitation (Ex): 390
- Storage conditions for powder: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light)
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light)
- Solubility in DMSO: 300 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.