Learn More
Carbosynth LLC. Ac-Leu-Arg-Ser-Arg-MCA, 5mg, MW: 729.84, Molecular formula: C33H51N11O8
SDP
Supplier:  Carbosynth LLC. MCA3952PI5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Ac-LRSR-AMC, Ac-Leu-Arg-Ser-Arg-AMC, Ac-LRSR-MCA, Ac-LRSR-MCA, Application: Substrate for MALT1, Storage Temperature: -20C, References: F. Rebeaud, S. Hailfinger, A. Posevitz-Fejfar, M. Tapernoux, R. Moser, D. Rueda, O. Gaide, M. Guzzardi, E.M. Iancu, N. Rufer, N. Fasel, and M. Thome, Nature Immunology, 9, 272 (2008).
Catalog No. 50-168-5967