Learn More
Enzo Life Sciences Ac-LEVD-CHO (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260065M001
Encompass_Preferred
Inhibitor of caspase-4 and -5. Reversible inhibitor of caspase-4. Also inhibits caspase-5. Alternative name: Caspase 4 inhibitor (aldehyde). Formula: C22H36N4O9. MW: 500.6. Purity: ≥95% (HPLC). Appearance: White powder. Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in distilled water. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8281