Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-LEVD-CHO | 402832-01-3 | 98.4% | 500.54 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY136727

Encompass_Preferred

Ac-LEVD-CHO is a caspase-4 inhibitor, a peptide with the sequence Ac-Leu-Glu-Val-Asp-al. It is provided as a solid.

  • Caspase-4 inhibitor
  • Peptide sequence Ac-Leu-Glu-Val-Asp-al
  • Purity of 98.4%
  • Appears as a white to off-white solid
  • Soluble in DMSO and water
  • Store powder at -80°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months

Catalog No. 50-003-48037


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.