Learn More
Cole-Parmer Extech 480172 3-Prong 120 VAC Line Separator
Supplier: Cole-Parmer 2684254
This AC line separator provides temporary separation of conductors for current measurement with clamp-type meters. Simply plug the separator into a grounded 120 VAC receptacle and then plug it in to your equipment. Turn it on and then measure.