Learn More
Bachem Ac-Lys(Ac)-D-Ala-D-Ala-OH
SDP
Supplier:  Bachem 4033018.0050
Encompass_Preferred
Ac-Lys(Ac)-D-Ala-D-Ala-OH, 24570-39-6, C16H28N4O6, Mr 372.42, 50mg. Diacetyl-Lys-D-Ala-D-Ala-OH, Ac2-Lys-D-Ala-D-Ala-OH, DALAA, N,N,Diacetyl-Lys-DAla-DAla. Diacetyl-Lys-D-Ala-D-Ala (DALAA) is a substrate for penicillin-sensitive D-alanine carboxypeptidases (DD-carboxypeptidases).
Catalog No. 50-260-450