Learn More
Bachem Ac-Lys-AMC
SDP
Supplier:  Bachem 4003625.0250
Encompass_Preferred
Ac-Lys-AMC Acetate, 156661-42-6, C18H23N3O4, Mr 345.40, 250mg. Ac-Lys-AMC is cleaved by trypsin.
Catalog No. 50-260-272