Learn More
Medchemexpress LLC Ac-Lys-Val-Cit-PABC- | 5mg
SDP
Supplier:  Medchemexpress LLC HY147286A5MG
Encompass_Preferred
Ac-Lys-Val-Cit-PABC-MMAE (formic) is a agent-linker conjugate for ADC Ac-Lys-Val-Cit-PABC-MMAE contains the ADC linker (peptide Ac-Lys-Val-Cit-PABC) and a potent tubulin polymerization inhibitorMonomethyl auristatin E (HY-15162)[1]
Catalog No. 50-002-02071