Learn More
Medchemexpress LLC Ac-lys-val-cit-pabc-mmae | 1650569-89-3
SDP
Supplier:  Medchemexpress LLC HY147286100MG
Encompass_Preferred
Ac-Lys-Val-Cit-PABC-MMAE is a agent-linker conjugate for ADC Ac-Lys-Val-Cit-PABC-MMAE contains the ADC linker (peptide Ac-Lys-Val-Cit-PABC) and a potent tubulin polymerization inhibitor MMAE (HY-15162)[1]
Catalog No. 50-002-03633