Learn More
Enzo Life Sciences Ac-muramyl-Ala-D-Glu-amide (5mg). CAS: 53678-77-6
SDP
Supplier:  Enzo Life Sciences ALX151035M005
Encompass_Preferred
NALP3 ligand. Specific ligand for NALP3. For inactive control compound see MDP-DD (ALX-151-036). Alternative name: MDP-LD, N-Acetylmuramyl-L-alanyl-D-isoglutamine, Adjuvant peptide. Formula: C19H32N4O11. MW: 492.5. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C. Handling: After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-7769