Learn More
Bachem Ac-muramyl-Ala-D-Glu-NH2
SDP
Supplier:  Bachem 4009623.0005
Encompass_Preferred
Ac-muramyl-Ala-D-Glu-NH2, 53678-77-6, C19H32N4O11, Mr 492.48, 5mg. MDP, Ac-muramyl-Ala-D-Isogln-OH, Muramyl Dipeptide, Adjuvant Peptide. Muramyl dipeptide (MDP) inhibits HIV replication in CD4+ H9 lymphocytes. .
Catalog No. 50-259-251