Learn More
Carbosynth LLC. Ac-Nle-Cyclo[Asp-His-D-Phe-Arg-Trp-Lys]-NH2, 1mg, MW: 1024.2, Molecular formula: C50H69N15O9, CAS# 121062-08-6
Supplier: Carbosynth LLC. PMC3683PI1MG
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Ac-Nle-cyclo[Asp-His-D-Phe-Arg-Trp-Lys]-NHsub>2; Ac-Nle-cyclo[DHfRWK]-NH2; Melanotan II; MT-II, Ac-Nle-cyclo [Asp-His-D-Phe-Arg-Trp-Lys]-NH2, Ac-Nle-c[DHfRWK]-NH2, Application: Potent Melanocortin 1, 3, 4 and 5 Receptor Agonist, Storage Temperature: -20C, References: F. Al-Obeidi,V.J. Hruby, A.M. Castrucci, and M.E. Hadley, J. Med. Chem., 32,174 (1989).