Learn More
Medchemexpress LLC Ac-Nle-Pro-Nle-Asp-AMC | 355140-49-7 | 98.7% | 655.74 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP20161MG
Encompass_Preferred
Ac-Nle-Pro-Nle-Asp-AMC is a specific substrate primarily utilized for analyzing the caspase-like activity of the 26S proteasome.
  • Specific substrate for 26S proteasome activity analysis.
  • Used to study caspase-like activity.
Catalog No. 50-004-24140