Learn More
Medchemexpress LLC Ac-Phe-Lys-OH | 14287-21-9 | 99.7% | 335.40 | 10 MG

Supplier: Medchemexpress LLC HYP484010MG
Ac-Phe-Lys-OH is an inverted Aspartame type sweetener.
- Molecular formula: C17H25N3O4
- Molecular weight: 335.40
- Sequence: Ac-Phe-Lys-OH (Ac-FK-OH)
- Purity: 99.7% by LCMS
- Recommended storage as powder: 2 years at -80°C or 1 year at -20°C in sealed, moisture-free conditions
- Recommended storage in solvent: 6 months at -80°C or 1 month at -20°C in sealed, moisture-free conditions
- Soluble in 1 M HCl and water (with adjustments)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.