Learn More
Medchemexpress LLC Ac-Phe-Lys-OH | 14287-21-9 | 99.7% | 335.40 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP4840
Ac-Phe-Lys-OH is a peptide (CAS 14287-21-9) with a molecular weight of 335.40 and a purity of 99.7%. This compound is recognized as an inverted Aspartame type sweetener and is intended for research use.
- High purity: 99.7%
- Molecular weight: 335.40
- Sealed storage, away from moisture
- Powder stable for up to 2 years at -80°C, or 1 year at -20°C
- In solvent stable for up to 6 months at -80°C, or 1 month at -20°C
- Functions as an inverted Aspartame type sweetener
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.