Learn More
Medchemexpress LLC Ac-pSar12-OH | 2313534-24-4 | 99.5% | 912.99 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1006910MG
Encompass_Preferred
Ac-pSar12-OH is a water-soluble polymer from the polysarcosine class, used as a linker. It is primarily applied in biopharmaceutical fields such as drug conjugation and nanoformulations.
  • Used in the synthesis of antibody-drug conjugates.
  • Can modify nanocarriers.
  • Can modify peptides and proteins.
Catalog No. 50-004-24046