Learn More
Cayman Chemical Ac-QQQ-AMChydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2771110 MG
Encompass_Preferred
A fluorogenic probe; QQQ is a substrate for peptidoglutaminase I and II from the bacterium B. circulans; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-7947