Learn More
CPC Scientific Ac-Trp-Glu-Ala-Asp-pNA (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CASP039A
Encompass
ONE-LETTER SEQUENCE: Ac-WEAD-pNA MOLECULAR FORMULA: C31H35N7O11MOLECULAR WEIGHT:681.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: D. Lee, et al., Bioorg. Med. Chem. Lett. 9, 1667 (1999); T.A. Rano, et al., Chem. Biol. 4, 149 (1997)
Catalog No. 50-210-8988