Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-Trp-Glu-His-Asp-AMC | 189275-74-9 | 99.8% | 784.77 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP45475MG

Encompass_Preferred

Ac-Trp-Glu-His-Asp-AMC (AC-WEHD-AMC) is a potent fluorogenic substrate of caspase-1. This product is for research use only and not intended for patient use.

  • Solid appearance
  • White to off-white color
  • Sequence: Ac-Trp-Glu-His-{Asp-AMC}
  • Sequence shortening: Ac-WEH-{Asp-AMC}
  • Shipping at room temperature in continental US; may vary elsewhere
  • Sealed storage, away from moisture for powder: -80°C for 2 years, -20°C for 1 year
  • Sealed storage, away from moisture for in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: ≥ 100 mg/mL (127.43 mM)

Catalog No. 50-003-97531


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.