Learn More
Medchemexpress LLC Ac-Trp-Glu-His-Asp-AMC (AC-WEHD-AMC) | 189275-74-9 | 99.84% | 784.77 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP45471MG
Encompass_Preferred
Ac-Trp-Glu-His-Asp-AMC (AC-WEHD-AMC) is a potent fluorogenic substrate of caspase-1. It is intended for research use only and is not sold to patients.
Catalog No. 50-003-96473